Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lact-7
Entrez Gene ID
183371
Gene Name
Beta-lactamase domain-containing protein
Gene Aliases
CELE_C40H5.4
Chromosome Location
Chr.X: 11759109 - 11764535 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077568.4 NP_509969.2 13 - 14 2016 69

View Full Product Details