Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttx-3
Entrez Gene ID
181357
Gene Name
protein-coding
Gene Aliases
CELE_C40H5.5
Chromosome Location
Chr.X: 11764456 - 11769378 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270239.2 NP_001257168.1 2 - 3 317 138
NM_001270240.3 NP_001257169.1 1 - 2 233 138
GenBank mRNA U72211.1 - 2 - 3 317 138

View Full Product Details