Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rsd-3
Entrez Gene ID
181358
Gene Name
ENTH domain-containing protein
Gene Aliases
CELE_C34E11.1
Chromosome Location
Chr.X: 11798505 - 11803656 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077572.8 NP_509973.1 9 - 10 1362 87

View Full Product Details