Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
F54F7.14
Entrez Gene ID
27307934
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F54F7.14
Chromosome Location
Chr.X: 11850872 - 11851735 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077578.3 NP_509979.2 1 - 2 165 165

View Full Product Details