Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tsp-20
Entrez Gene ID
181376
Gene Name
TetraSPanin family
Gene Aliases
CELE_B0198.1
Chromosome Location
Chr.X: 12031592 - 12034037 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077613.7 NP_510014.1 1 - 2 128 82

View Full Product Details