Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-24
Entrez Gene ID
6418837
Gene Name
Neur_chan_LBD domain-containing protein
Gene Aliases
CELE_C15A7.4
Chromosome Location
Chr.X: 12068550 - 12070563 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129638.3 NP_001123110.2 4 - 5 553 129

View Full Product Details