Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-15
Entrez Gene ID
187973
Gene Name
Neur_chan_LBD domain-containing protein
Gene Aliases
CELE_T01H10.3
Chromosome Location
Chr.X: 12114324 - 12115923 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077624.2 NP_510025.2 5 - 6 661 77

View Full Product Details