Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lgc-18
Entrez Gene ID
187977
Gene Name
Neur_chan_LBD domain-containing protein
Gene Aliases
CELE_T01H10.7
Chromosome Location
Chr.X: 12125608 - 12128000 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077628.3 NP_510029.3 5 - 6 655 147

View Full Product Details