Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-45
Entrez Gene ID
184603
Gene Name
Serpentine receptor class delta-45
Gene Aliases
CELE_F17A2.7
Chromosome Location
Chr.X: 12328976 - 12330526 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077663.1 NP_510064.1 1 - 2 284 150

View Full Product Details