Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-48
Entrez Gene ID
184605
Gene Name
Serpentine receptor class delta-48
Gene Aliases
CELE_F17A2.9
Chromosome Location
Chr.X: 12336108 - 12337752 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077666.1 NP_510067.1 5 - 6 785 123

View Full Product Details