Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-27
Entrez Gene ID
41656994
Gene Name
protein-coding
Gene Aliases
CELE_F16H9.2
Chromosome Location
Chr.X: 12705624 - 12808318 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270268.1 NP_001257197.1 2 - 3 118 75
NM_001381126.1 NP_001366722.1 2 - 3 187 75

View Full Product Details