Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pgp-12
Entrez Gene ID
181415
Gene Name
P-GlycoProtein related
Gene Aliases
CELE_F22E10.1
Chromosome Location
Chr.X: 12731807 - 12737441 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077725.6 NP_510126.1 8 - 9 1225 89

View Full Product Details