Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
chd-3
Entrez Gene ID
181421
Gene Name
Chromodomain-helicase-DNA-binding protein 3 homolog
Gene Aliases
CELE_T14G8.1
Chromosome Location
Chr.X: 12843873 - 12852601 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077739.7 NP_510140.1 3 - 4 1808 80
GenBank mRNA AF308444.1 - 3 - 4 1830 80

View Full Product Details