Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpx-2
Entrez Gene ID
186917
Gene Name
ComPleXin (synaptic protein) homolog
Gene Aliases
CELE_K03A11.2
Chromosome Location
Chr.X: 13066974 - 13071808 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383662.1 4 - 5 486 96

View Full Product Details