Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cwp-5
Entrez Gene ID
185977
Gene Name
protein-coding
Gene Aliases
CELE_F48C11.2
Chromosome Location
Chr.X: 13182114 - 13189961 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373548.2 NP_001359731.1 7 - 8 1559 141
NM_001373549.3 NP_001359732.1 5 - 6 1208 141

View Full Product Details