Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpa-6
Entrez Gene ID
181442
Gene Name
Guanine nucleotide-binding protein alpha-6 subunit
Gene Aliases
CELE_F48C11.1
Chromosome Location
Chr.X: 13191212 - 13194390 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077788.2 NP_510189.1 8 - 9 1063 84
GenBank mRNA AY008129.1 - 8 - 9 1029 84

View Full Product Details