Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-14A1
Entrez Gene ID
187183
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_K09A11.2
Chromosome Location
Chr.X: 13265041 - 13267359 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077802.4 NP_510203.1 6 - 7 1289 96

View Full Product Details