Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-14A3
Entrez Gene ID
187185
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_K09A11.4
Chromosome Location
Chr.X: 13271837 - 13274218 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077804.3 NP_510205.1 5 - 6 1123 145

View Full Product Details