Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-36
Entrez Gene ID
187574
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_R04D3.6
Chromosome Location
Chr.X: 13293361 - 13294773 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077811.3 NP_510212.1 1 - 2 303 109

View Full Product Details