Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
flr-1
Entrez Gene ID
181468
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F02D10.5
Chromosome Location
Chr.X: 13452667 - 13456658 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077842.4 NP_510243.1 8 - 9 961 69
GenBank mRNA AB012617.1 - 8 - 9 867 69

View Full Product Details