Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acr-12
Entrez Gene ID
181475
Gene Name
protein-coding
Gene Aliases
CELE_R01E6.4
Chromosome Location
Chr.X: 13532853 - 13536459 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313441.4 NP_001300370.1 1 - 2 113 81
NM_077861.6 NP_510262.1 3 - 4 576 81
GenBank mRNA AF187013.1 - 3 - 4 527 81

View Full Product Details