Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-20
Entrez Gene ID
187506
Gene Name
GLoBin related
Gene Aliases
CELE_R01E6.6
Chromosome Location
Chr.X: 13552339 - 13556166 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077866.4 NP_510267.2 2 - 3 254 84

View Full Product Details