Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-9
Entrez Gene ID
186210
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_F54B11.1
Chromosome Location
Chr.X: 13590035 - 13591207 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077872.1 NP_510273.1 1 - 2 165 95

View Full Product Details