Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-20
Entrez Gene ID
181485
Gene Name
Two pore potassium channel protein sup-9
Gene Aliases
CELE_C40C9.1
Chromosome Location
Chr.X: 13639877 - 13642359 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077883.2 NP_510284.1 7 - 8 611 79
GenBank mRNA AF083646.1 - 7 - 8 611 79

View Full Product Details