Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acr-9
Entrez Gene ID
191597
Gene Name
AcetylCholine Receptor
Gene Aliases
CELE_C40C9.2
Chromosome Location
Chr.X: 13642313 - 13646898 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077884.5 NP_510285.2 10 - 11 1387 111

View Full Product Details