Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
meg-2
Entrez Gene ID
181501
Gene Name
Uncharacterized protein
Gene Aliases
CELE_K02B9.2
Chromosome Location
Chr.X: 13922643 - 13926129 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077917.5 NP_510318.2 2 - 3 462 61

View Full Product Details