Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-39
Entrez Gene ID
181506
Gene Name
Prion-like-(Q/N-rich)-domain-bearing protein
Gene Aliases
CELE_F48F7.4
Chromosome Location
Chr.X: 13965676 - 13968676 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077924.5 NP_510325.2 6 - 7 1359 85

View Full Product Details