Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbc-19
Entrez Gene ID
181511
Gene Name
TBC (Tre-2/Bub2/Cdc16) domain family
Gene Aliases
CELE_C31E10.8
Chromosome Location
Chr.X: 14003020 - 14012595 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077935.7 NP_510336.3 21 - 22 3162 143

View Full Product Details