Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-25
Entrez Gene ID
181525
Gene Name
Neur_chan_LBD domain-containing protein
Gene Aliases
CELE_R03E1.3
Chromosome Location
Chr.X: 14158785 - 14161734 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077958.6 NP_510359.3 8 - 9 1248 82

View Full Product Details