Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sym-4
Entrez Gene ID
181527
Gene Name
Uncharacterized protein
Gene Aliases
CELE_R03E1.1
Chromosome Location
Chr.X: 14164569 - 14170746 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077960.7 NP_510361.1 4 - 5 548 87
GenBank mRNA AY372076.1 - 2 - 3 339 87

View Full Product Details