Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sym-4
Entrez Gene ID
181527
Gene Name
Uncharacterized protein
Gene Aliases
CELE_R03E1.1
Chromosome Location
Chr.X: 14164569 - 14170746 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077960.7 NP_510361.1 7 - 8 1807 65
GenBank mRNA AY372076.1 - 5 - 6 1598 65

View Full Product Details