Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-13B2
Entrez Gene ID
187068
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_K06G5.2
Chromosome Location
Chr.X: 14225573 - 14228785 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077968.6 NP_510369.2 6 - 7 820 116

View Full Product Details