The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001368544.2 | NP_001355359.1 | 12 - 13 | 1512 | 116 |
NM_001368545.3 | NP_001355470.1 | 13 - 13 | 1656 | 116 | |
NM_001368546.2 | NP_001355471.1 | 12 - 13 | 1461 | 116 |