Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
del-1
Entrez Gene ID
181534
Gene Name
protein-coding
Gene Aliases
CELE_E02H4.1
Chromosome Location
Chr.X: 14268081 - 14277082 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001368544.2 NP_001355359.1 13 - 14 1624 78
NM_001368545.3 NP_001355470.1 14 - 14 1768 78
NM_001368546.2 NP_001355471.1 13 - 14 1573 78

View Full Product Details