Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sel-7
Entrez Gene ID
181539
Gene Name
Suppressor/Enhancer of Lin-12
Gene Aliases
CELE_K04G11.2
Chromosome Location
Chr.X: 14332738 - 14340481 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077991.7 NP_510392.2 6 - 7 809 121
GenBank mRNA AY398504.1 - 6 - 7 800 121

View Full Product Details