Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sym-1
Entrez Gene ID
181555
Gene Name
Uncharacterized protein
Gene Aliases
CELE_C44H4.3
Chromosome Location
Chr.X: 14581622 - 14584378 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078025.7 NP_510426.1 4 - 5 828 93
GenBank mRNA AJ242473.1 - 4 - 5 732 93

View Full Product Details