Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hcp-3
Entrez Gene ID
176359
Gene Name
Histone H3-like centromeric protein hcp-3
Gene Aliases
CELE_F58A4.3
Chromosome Location
Chr.III: 9615306 - 9616470 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066727.7 NP_499128.1 3 - 4 659 126

View Full Product Details