Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ent-1
Entrez Gene ID
178097
Gene Name
protein-coding
Gene Aliases
CELE_ZK809.4
Chromosome Location
Chr.IV: 11653417 - 11655470 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268644.2 NP_001255573.1 5 - 6 1035 135
NM_001268645.3 NP_001255574.1 1 - 2 81 135
GenBank mRNA AF435979.1 - 5 - 6 1029 135

View Full Product Details