Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scrm-5
Entrez Gene ID
177180
Gene Name
Phospholipid scramblase
Gene Aliases
CELE_K08D10.8
Chromosome Location
Chr.IV: 4162421 - 4164480 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068099.7 NP_500500.1 4 - 5 815 175

View Full Product Details