Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-26
Entrez Gene ID
187979
Gene Name
Serpentine receptor class delta-26
Gene Aliases
CELE_T02B5.4
Chromosome Location
Chr.V: 14161459 - 14163009 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074104.2 NP_506505.1 2 - 3 831 159

View Full Product Details