Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-49
Entrez Gene ID
186204
Gene Name
protein-coding
Gene Aliases
CELE_F54B8.7
Chromosome Location
Chr.V: 15821698 - 15822884 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028795.1 NP_001023966.1 2 - 3 637 125
NM_001028796.1 NP_001023967.1 2 - 3 406 125

View Full Product Details