Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- fipr-23
- Entrez Gene ID
-
173283
- Gene Name
- FIP (Fungus-Induced Protein) Related
- Gene Aliases
-
CELE_C37A5.4
- Chromosome Location
-
Chr.I: 14153228 - 14154195 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061068.5 |
NP_493469.1 |
1 - 2 |
261 |
143 |
- Gene Symbol
- C37A5.3
- Entrez Gene ID
-
13182956
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_C37A5.3
- Chromosome Location
-
Chr.I: 14153228 - 14155296 on Build WBcel235
View Full Product Details