Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-73
Entrez Gene ID
184366
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_F11G11.12
Chromosome Location
Chr.II: 4872098 - 4873129 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062479.7 NP_494880.1 1 - 2 813 125

View Full Product Details