Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-13A5
Entrez Gene ID
188356
Gene Name
Putative cytochrome P450 CYP13A5
Gene Aliases
CELE_T10B9.2
Chromosome Location
Chr.II: 9798277 - 9800413 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063711.5 NP_496112.1 4 - 5 1253 91

View Full Product Details