Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxb-84
Entrez Gene ID
188568
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_T17A3.7
Chromosome Location
Chr.III: 142093 - 146482 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064756.3 NP_497157.1 1 - 2 95 121

View Full Product Details