Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-51
Entrez Gene ID
188440
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_T12B5.1
Chromosome Location
Chr.III: 962269 - 963833 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064906.2 NP_497307.1 2 - 3 768 110

View Full Product Details