Assay Design
The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Gene Transcripts
- Gene Symbol
- rpl-29
- Entrez Gene ID
-
178352
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_B0513.3
- Chromosome Location
-
Chr.IV: 13882225 - 13882618 on Build WBcel235
- Gene Symbol
- thoc-7
- Entrez Gene ID
-
178353
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_B0513.2
- Chromosome Location
-
Chr.IV: 13882219 - 13884192 on Build WBcel235
View Full Product Details