Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fip-7
Entrez Gene ID
178461
Gene Name
Fungus-Induced Protein
Gene Aliases
CELE_Y51H4A.10
Chromosome Location
Chr.IV: 16626205 - 16631251 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383259.1 2 - 2 80 79

View Full Product Details