Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lec-8
Entrez Gene ID
181198
Gene Name
Probable galaptin lec-8
Gene Aliases
CELE_R07B1.10
Chromosome Location
Chr.X: 9850209 - 9851336 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077248.5 NP_509649.1 3 - 3 902 71
GenBank mRNA AB038499.1 - 3 - 3 521 71

View Full Product Details