Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srab-24
Entrez Gene ID
3564812
Gene Name
Serpentine Receptor, class AB (class A-like)
Gene Aliases
CELE_Y64G10A.8
Chromosome Location
Chr.IV: 14245418 - 14250213 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028376.2 NP_001023547.1 3 - 4 455 75

View Full Product Details